Polyethylene glycol 6,000
Supplier: TCI America
P0903-25G
P0903-500G
TCP0903-25GEA
47.29
CAD
TCP0903-25G
TCP0903-500G
Polyethylene glycol 6,000
Polyethylene glycol 6,000
CAS Number: 25322-68-3
MDL Number: MFCD01779614
Form: Crystal
Color: White
Melting point (°C): 62
Flash Point (°C): 270
Other Product Names / Synonyms: PEG 6,000
Formula:
H(OCH₂CH₂)nOH Boiling Pt: ∼250 °C (1013 hPa) Melting Pt: 62 °C Density: 1.13 g/cm³ (20 °C) Flash Pt: 270 °C Storage Temperature: Ambient |
MDL Number:
MFCD01779614 CAS Number: 25322-68-3 EINECS: 500-038-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...