Propylene glycol diacetate ≥98.0%

Supplier: TCI America

P0902-25ML P0902-500ML
TCP0902-025MLEA 26.83 CAD
TCP0902-025ML TCP0902-500ML
Propylene glycol diacetate ≥98.0%
Propylene glycol diacetate

CAS Number: 623-84-7
MDL Number: MFCD00026201
Molecular Formula: C7H12O4
Molecular Weight: 160.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 186
Melting point (°C): -31
Flash Point (°C): 89
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: 1,2-Propanediol Diacetate, Propylene Diacetate, Propylene Glycol Diacetate

Formula: C₇H₁₂O₄
Boiling Pt: 186 °C
Melting Pt: –31 °C
Density: 1.06 (20 °C)
Flash Pt: 89 °C
MDL Number: MFCD00026201
CAS Number: 623-84-7
EINECS: 210-817-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=125a653de4b6b152ec30fab5cb7b1514ed3fbf5a&vid=125a653de4b6b152ec30fab5cb7b1514ed3fbf5a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR