4-Phenylimidazole ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Phenyl-1H-imidazole

P0877-25G P0877-5G
TCP0877-25GEA 509.08 CAD
TCP0877-25G TCP0877-005G
4-Phenylimidazole ≥98.0% (by titrimetric analysis)
4-Phenylimidazole

CAS Number: 670-95-1
MDL Number: MFCD00005197
Molecular Formula: C9H8N2
Molecular Weight: 144.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 146

Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 182…184 °C (1 mmHg)
Melting Pt: 146 °C
MDL Number: MFCD00005197
CAS Number: 670-95-1
EINECS: 211-580-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5de462cab8f5c46416e6171daf96125534d7af5f&vid=5de462cab8f5c46416e6171daf96125534d7af5f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR