Phenyltrimethoxysilane ≥98.0%

Supplier: TCI America

P0857-25ML P0857-500ML P0857-100ML
TCP0857-025MLEA 28.44 CAD
TCP0857-025ML TCP0857-500ML TCP0857-100ML
Phenyltrimethoxysilane ≥98.0%
Phenyltrimethoxysilane

CAS Number: 2996-92-1
MDL Number: MFCD00025689
Molecular Formula: C9H14O3Si
Molecular Weight: 198.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 131
Melting point (°C): -25
Flash Point (°C): 94
Specific Gravity (20/20): 1.07


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Phenyltrimethoxysilane

Formula: C₉H₁₄O₃Si
Boiling Pt: 131 °C
Melting Pt: –25 °C
Density: 1.07 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD00025689
CAS Number: 2996-92-1
EINECS: 221-066-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9493bb4429e3e01bd6bd74121c6058a8553b387&vid=d9493bb4429e3e01bd6bd74121c6058a8553b387&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR