2,3,4,5,6-Pentafluorobenzyl alcohol ≥96.0%
Supplier: TCI America
|
Warning
|
P0849-5G
P0849-25G
TCP0849-005GEA
120.08
CAD
TCP0849-005G
TCP0849-025G
2,3,4,5,6-Pentafluorobenzyl alcohol ≥96.0%
2,3,4,5,6-Pentafluorobenzyl alcohol
CAS Number: 440-60-8
MDL Number: MFCD00004602
Molecular Formula: C7H3F5O
Molecular Weight: 198.09
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 182
Melting point (°C): 37
Flash Point (°C): 87
Formula:
C₇H₃F₅O Boiling Pt: 182 °C Melting Pt: 37 °C Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004602 CAS Number: 440-60-8 EINECS: 207-126-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...