Perfluorohexane ≥96.0%
Supplier: TCI America
|
Warning
|
P0839-250G
P0839-25G
TCP0839-250GEA
2321.02
CAD
TCP0839-250G
TCP0839-25G
Perfluorohexane ≥96.0%
Perfluorohexane
CAS Number: 355-42-0
MDL Number: MFCD00000437
Molecular Formula: C6F14
Molecular Weight: 338.04
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 60
Specific Gravity (20/20): 1.69
Product Categories: Synthetic Organic Chemistry, Fluorous Chemistry
Other Product Names / Synonyms: Perfluorohexane
Formula:
C₆F₁₄ Boiling Pt: 60 °C Melting Pt: –4 °C Density: 1.69 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000437 CAS Number: 355-42-0 EINECS: 206-585-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...