Pivalamide ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

P0819-25G P0819-500G P0819-100G
TCP0819-025GEA 67.78 CAD
TCP0819-025G TCP0819-500G TCP0819-100G
Pivalamide ≥98.0% (by GC, total nitrogen)
Pivalamide

CAS Number: 754-10-9
MDL Number: MFCD00008011
Molecular Formula: C5H11NO
Molecular Weight: 101.15
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Boiling point (°C): 212
Melting point (°C): 154


Other Product Names / Synonyms: 2,2-Dimethylpropionamide, Trimethylacetamide

Formula: C₅H₁₁NO
Boiling Pt: 212 °C
Melting Pt: 154 °C
Density: 1.02 g/cm³ (20 °C)
MDL Number: MFCD00008011
CAS Number: 754-10-9
EINECS: 212-043-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57a20f6ca7944adc3bcd88f95b204338c6f33dc4&vid=57a20f6ca7944adc3bcd88f95b204338c6f33dc4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR