2-Pentyn-1-ol ≥98.0%
Supplier: TCI America
|
Warning
|
P0815-25ML
P0815-5ML
TCP0815-025MLEA
308.1
CAD
TCP0815-025ML
TCP0815-005ML
2-Pentyn-1-ol ≥98.0%
2-Pentyn-1-ol
CAS Number: 6261-22-9
MDL Number: MFCD00040915
Molecular Formula: C5H8O
Molecular Weight: 84.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 154
Flash Point (°C): 62
Specific Gravity (20/20): 0.91
Formula:
C₅H₈O Boiling Pt: 154 °C Melting Pt: –0.2 °C Density: 0.91 (20 °C) Flash Pt: 62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040915 CAS Number: 6261-22-9 EINECS: 228-411-2 UN: 1987 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...