1-(2-Hydroxyethyl)piperazine ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
N-(2-Hydroxyethyl)piperazine, 2-(1-Piperazinyl)ethanol, 2-Piperazin-1-ylethanol
P0812-25G
P0812-500G
P0812-100G
TCP0812-025GEA
30.56
CAD
TCP0812-025G
TCP0812-500G
TCP0812-100G
1-(2-Hydroxyethyl)piperazine ≥99.0%
1-(2-Hydroxyethyl)piperazine
CAS Number: 103-76-4
MDL Number: MFCD00005970
Molecular Formula: C6H14N2O
Molecular Weight: 130.19
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 246
Flash Point (°C): 124
Specific Gravity (20/20): 1.06
Other Product Names / Synonyms: 1-Piperazineethanol, 2-(1-Piperazinyl)ethanol
Formula:
C₆H₁₄N₂O MW: 130.19 g/mol Boiling Pt: 246 °C Melting Pt: –38 °C Density: 1.06 (20 °C) Flash Pt: 124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005970 CAS Number: 103-76-4 EINECS: 203-142-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...