Pentafluorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

P0806-25G P0806-5G
TCP0806-025GEA 110.76 CAD
TCP0806-025G TCP0806-5G
Pentafluorobenzoic acid ≥98.0% (by GC, titration analysis)
Pentafluorobenzoic acid

CAS Number: 602-94-8
MDL Number: MFCD00002406
Molecular Formula: C7HF5O2
Molecular Weight: 212.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 104


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Perfluorobenzoic Acid

Formula: C₇HF₅O₂
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 104 °C
Density: 1.942 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002406
CAS Number: 602-94-8
EINECS: 210-026-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27f15adafe7dea175d110fa4db1060bb35016ca6&vid=4eb4908389940af672589c44018e379c801e2773&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR