2,6-Pyridinedimethanol ≥98.0%

Supplier: TCI America

Synonyms: 2,6-Bis(hydroxymethyl)pyridine

P0803-5G P0803-25G
TCP0803-005GEA 94.8 CAD
TCP0803-005G TCP0803-25G
2,6-Pyridinedimethanol ≥98.0%
2,6-Pyridinedimethanol

CAS Number: 1195-59-1
MDL Number: MFCD00006351
Molecular Formula: C7H9NO2
Molecular Weight: 139.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 185
Melting point (°C): 113
Flash Point (°C): 194


Other Product Names / Synonyms: 2,6-Bis(hydroxymethyl)pyridine

Formula: C₇H₉NO₂
MW: 139.15 g/mol
Boiling Pt: 185 °C
Melting Pt: 113 °C
Flash Pt: 194 °C
MDL Number: MFCD00006351
CAS Number: 1195-59-1
EINECS: 214-803-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa876843e1e9f33f063cc46f76fff0ef580acd3c&vid=fa876843e1e9f33f063cc46f76fff0ef580acd3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR