1,3-Propanedithiol ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1,3-Dimercaptopropane

P0763-25ML P0763-100ML
TCP0763-025MLEA 130.19 CAD
TCP0763-025ML TCP0763-100ML
1,3-Propanedithiol ≥97.0% (by GC, titration analysis)
1,3-Propanedithiol

CAS Number: 109-80-8
MDL Number: MFCD00004904
Molecular Formula: C3H8S2
Molecular Weight: 108.22
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 170
Melting point (°C): -79
Flash Point (°C): 61
Specific Gravity (20/20): 1.08


Product Categories: Synthetic Organic Chemistry, Thiols, Protection & Derivatization Reagents (for Synthesis), Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,3-Dimercaptopropane, Trimethylene Dithiol

Formula: C₃H₈S₂
MW: 108.23 g/mol
Boiling Pt: 170 °C
Melting Pt: –79 °C
Density: 1.08 (20 °C)
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00004904
CAS Number: 109-80-8
EINECS: 203-706-9
UN: 2810
ADR: 6.1,III
Merck Index: 12,07984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45c4d1c5ffab5e8b33b5d2934d6355f4898842bc&vid=45c4d1c5ffab5e8b33b5d2934d6355f4898842bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR