Phenyltriethoxysilane ≥99.0%

Supplier: TCI America

P0735-500ML P0735-25ML P0735-100ML
TCP0735-500MLEA 189.6 CAD
TCP0735-500ML TCP0735-25ML TCP0735-100ML
Phenyltriethoxysilane ≥99.0%
Phenyltriethoxysilane

CAS Number: 780-69-8
MDL Number: MFCD00009065
Molecular Formula: C12H20O3Si
Molecular Weight: 240.37
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 235
Flash Point (°C): 111
Specific Gravity (20/20): 0.99


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Phenyltriethoxysilane

Formula: C₁₂H₂₀O₃Si
Boiling Pt: 235 °C
Melting Pt: 41 °C
Density: 0.99 (20 °C)
Flash Pt: 111 °C
MDL Number: MFCD00009065
CAS Number: 780-69-8
EINECS: 212-305-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2260d5b31b13e0f6ccd91c67ab5d50137c085af0&vid=2260d5b31b13e0f6ccd91c67ab5d50137c085af0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR