(±)-1-Phenyl-1,2-ethanediol ≥98.0%

Supplier: TCI America

P0686-25G P0686-250G
TCP0686-025GEA 162.74 CAD
TCP0686-025G TCP0686-250G
(±)-1-Phenyl-1,2-ethanediol ≥98.0%
(±)-1-Phenyl-1,2-ethanediol

CAS Number: 93-56-1
MDL Number: MFCD00003546
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 82
Melting point (°C): 67
Flash Point (°C): 160


Other Product Names / Synonyms: Styrene Glycol, Phenylethylene Glycol

Formula: C₆H₅CH(OH)CH₂OH
Boiling Pt: 82 °C
Melting Pt: 67 °C
Flash Pt: 160 °C
MDL Number: MFCD00003546
CAS Number: 93-56-1
EINECS: 202-258-1
Merck Index: 12,09029
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eac30ac4d73a63ad627939278dd241035186523f&vid=eac30ac4d73a63ad627939278dd241035186523f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR