Pyridine-3,4-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
P0682-5G
P0682-25G
TCP0682-005GEA
52.93
CAD
TCP0682-005G
TCP0682-025G
Pyridine-3,4-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Pyridine-3,4-dicarboxylic acid
CAS Number: 490-11-9
MDL Number: MFCD00006392
Molecular Formula: C7H5NO4
Molecular Weight: 167.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 260
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Cinchomeronic Acid
Formula:
C₇H₅NO₄ Boiling Pt: 529 °C (1013 hPa) Melting Pt: 260 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006392 CAS Number: 490-11-9 EINECS: 207-705-4 Merck Index: 13,02304 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...