Isonicotinaldehyde oxime ≥98.0%

Supplier: TCI America

Synonyms: 4-Pyridinecarbaldehyde oxime, Pyridine-4-carboxaldoxime

P0673-10G P0673-25G
TCP0673-010GEA 140.62 CAD
TCP0673-010G TCP0673-025G
Isonicotinaldehyde oxime ≥98.0%
Isonicotinaldehyde oxime

CAS Number: 696-54-8
MDL Number: MFCD00006446
Molecular Formula: C6H6N2O
Molecular Weight: 122.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 133
Flash Point (°C): 110

Formula: C₆H₆N₂O
Melting Pt: 133 °C
Flash Pt: 110 °C
MDL Number: MFCD00006446
CAS Number: 696-54-8
EINECS: 211-796-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b404f8ea90dae86269681630e98ab61978717fe6&vid=b404f8ea90dae86269681630e98ab61978717fe6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR