2,4-Pentanediol ≥98.0%
Supplier: TCI America
Synonyms:
2,4-Dihydroxy pentane
P0670-500G
P0670-25G
TCP0670-500GEA
1153.4
CAD
TCP0670-500G
TCP0670-025G
2,4-Pentanediol ≥98.0%
2,4-Pentanediol
CAS Number: 625-69-4
MDL Number: MFCD00004549
Molecular Formula: C5H12O2
Molecular Weight: 104.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 198
Flash Point (°C): 101
Specific Gravity (20/20): 0.96
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,4-Dihydroxypentane
Formula:
C₅H₁₂O₂ MW: 104.15 g/mol Boiling Pt: 198 °C Melting Pt: ∼53 °C Density: 0.96 (20 °C) Flash Pt: 101 Storage Temperature: Ambient |
MDL Number:
MFCD00004549 CAS Number: 625-69-4 EINECS: 210-907-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...