Pyridazine ≥99.0%
Supplier: TCI America
P0616-5G
P0616-25G
TCP0616-005GEA
94.8
CAD
TCP0616-005G
TCP0616-025G
Pyridazine ≥99.0%
Pyridazine
CAS Number: 289-80-5
MDL Number: MFCD00006463
Molecular Formula: C4H4N2
Molecular Weight: 80.09
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Melting point (°C): -8
Flash Point (°C): 85
Specific Gravity (20/20): 1.11
Formula:
C₄H₄N₂ Boiling Pt: 208 °C Melting Pt: –8 °C Density: 1.11 (20 °C) Flash Pt: 85 °C |
MDL Number:
MFCD00006463 CAS Number: 289-80-5 EINECS: 206-025-5 Merck Index: 12,08152 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...