Phenylpropiolic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Phenylacetylenecarboxylic acid, Phenylpropynoic acid, 3-Phenylpropiolic acid

P0610-5G P0610-25G
TCP0610-005GEA 91.09 CAD
TCP0610-005G TCP0610-025G
Phenylpropiolic acid ≥98.0% (by titrimetric analysis)
Phenylpropiolic acid

CAS Number: 637-44-5
MDL Number: MFCD00004361
Molecular Formula: C9H6O2
Molecular Weight: 146.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 137


Product Categories: Synthetic Organic Chemistry, Carboxylic Acids (Multi-component Reaction)
Other Product Names / Synonyms: Phenylpropargylic Acid

Formula: C₉H₆O₂
MW: 146.15 g/mol
Boiling Pt: 294 °C (1013 hPa)
Melting Pt: 137 °C
Density: 1.28 g/cm³ (20 °C)
MDL Number: MFCD00004361
CAS Number: 637-44-5
EINECS: 211-285-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb2406578d2aef09fda849828371685b915418b4&vid=fb2406578d2aef09fda849828371685b915418b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR