n-Pentadecane ≥99.0%
Supplier: TCI America
|
Danger
|
P0606-500ML
P0606-25ML
P0606-100ML
TCP0606-500MLEA
510.34
CAD
TCP0606-500ML
TCP0606-025ML
TCP0606-100ML
n-Pentadecane ≥99.0%
n-Pentadecane
CAS Number: 629-62-9
MDL Number: MFCD00008990
Molecular Formula: C15H32
Molecular Weight: 212.42
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 270
Flash Point (°C): 120
Freezing point (°C): 10
Specific Gravity (20/20): 0.77
Formula:
H₃C(CH₂)₁₃CH₃ Boiling Pt: 270 °C Melting Pt: 13…15 °C Density: 0.77 (20 °C) Flash Pt: 120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008990 CAS Number: 629-62-9 EINECS: 211-098-1 UN: 3295 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...