Pararosaniline hydrochloride ≥95.0%

Supplier: TCI America
Danger

Synonyms: Basic Parafuchsin, 4,4'-(4-Iminocyclohexa-2,5-dienylidenemethylene)dianiline hydrochloride, [4-[Bis(4-aminophenyl)methylidene]-1-cyclohexa-2,5-dienylidene]azanium chloride, Basic Fuchsin hydrochloride, Parafuchsin hydrochloride, Pararosaniline chloride, Basic Red 9, 4,4'-((4-Iminocyclohexa-2,5-dien-1-ylidene)methylene)dianiline hydrochloride, Paramagenta hydrochloride

P0599-25G P0599-500G P0599-100G
TCP0599-025GEA 163.2 CAD
TCP0599-025G TCP0599-500G TCP0599-100G
Pararosaniline hydrochloride ≥95.0%
Pararosaniline hydrochloride

CAS Number: 569-61-9 MDL Number: MFCD00001657 Molecular Formula: C19H17N3 Molecular Weight: 323.82 Purity/Analysis Method: 95.0% (HPLC) Form: Crystal Melting point (°C): 270 Lambda max.: 539 nm (EtOH)


Product Categories: Bioscience, Stains & Dyes
Other Product Names / Synonyms: Parafuchsin, Basic Red 9

Formula: C₁₉H₁₈ClN₃
MW: 323.82 g/mol
Melting Pt: 270 °C
Storage Temperature: Ambient
MDL Number: MFCD00001657
CAS Number: 569-61-9
EINECS: 209-321-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca8dce159466d1c296acb284f5a64213ba64a3b1&vid=ca8dce159466d1c296acb284f5a64213ba64a3b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR