Sodium pyruvate ≥97.0%

Supplier: TCI America

Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt

P0582-25G P0582-500G
TCP0582-025GEA 54.51 CAD
TCP0582-025G TCP0582-500G
Sodium pyruvate ≥97.0%
Sodium pyruvate

CAS Number: 113-24-6
MDL Number: MFCD00002586
Molecular Formula: C3H4O3
Molecular Weight: 110.04
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Bioscience, Regenerative Medicine Research Reagents
Other Product Names / Synonyms: Pyruvic Acid Sodium Salt

Formula: CH₃COCOONa
Melting Pt: 220…230 °C
Density: 1.718 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-024-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=018f2387d02c63510aa375e1cc21ee99502228ea&vid=018f2387d02c63510aa375e1cc21ee99502228ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR