Pyrrole ≥99.0%
Supplier: TCI America
Synonyms:
1H-Pyrrole
P0574-500ML
P0574-100ML
P0574-25ML
TCP0574-100MLEA
161.89
CAD
TCP0574-25ML
TCP0574-100ML
TCP0574-500ML
Pyrrole ≥99.0%
Pyrrole
CAS Number: 109-97-7
MDL Number: MFCD00005216
Molecular Formula: C4H5N
Molecular Weight: 67.09
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Melting point (°C): -23
Flash Point (°C): 39
Specific Gravity (20/20): 0.97
Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: Divinylenimine
Formula:
C₄H₅N MW: 67.09 g/mol Boiling Pt: 130 °C Melting Pt: –23 °C Density: 0.97 (20 °C) Flash Pt: 39 °C |
MDL Number:
MFCD00005216 CAS Number: 109-97-7 UN: 1992 ADR: 3,III Merck Index: 12,08198 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...