(S)-(-)-2-Pyrrolidinone-5-carboxylic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (S)-(-)-2-Pyrrolidone-5-carboxylic acidL-Pyroglutamic Acid(S)-5-Oxopyrrolidine-2-carboxylic acidH-Pyr-OH

P0573-500G P0573-25G P0573-100G
TCP0573-100GEA 107.93 CAD
TCP0573-500G TCP0573-025G TCP0573-100G
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid ≥97.0% (by titrimetric analysis)
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid

CAS Number: 98-79-3
MDL Number: MFCD00005272
Molecular Formula: C5H7NO3
Molecular Weight: 129.12
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 160
Specific rotation [a]20/D: -10 deg (C=5, H2O)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Bioscience, Biologically-modified Amino Acids
Other Product Names / Synonyms: L-Glutamic Acid Lactam, (S)-5-Oxopyrrolidine-2-carboxylic Acid, (S)-2-Pyrrolidone-5-carboxylic Acid, H-Pyr-OH

Formula: C₅H₇NO₃
MW: 129.1 g/mol
Melting Pt: 160 °C
MDL Number: MFCD00005272
CAS Number: 98-79-3
https://ca.vwr.com/stibo/web/std.lang.all/13/02/28991302.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR