2,3,4-Trihydroxybenzoic acid ≥98.0%

Supplier: TCI America
Warning

P0571-1G P0571-5G
TCP0571-001GEA 34.76 CAD
TCP0571-001G TCP0571-005G
2,3,4-Trihydroxybenzoic acid ≥98.0%
2,3,4-Trihydroxybenzoic acid

CAS Number: 610-02-6
MDL Number: MFCD00002447
Molecular Formula: C7H6O5
Molecular Weight: 170.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Pyrogallol-4-carboxylic Acid

Formula: C₇H₆O₅
Storage Temperature: Ambient
MDL Number: MFCD00002447
CAS Number: 610-02-6
EINECS: 210-205-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8aa286be48f1797a4ef8429f261fe165d3c6bc4&vid=d8aa286be48f1797a4ef8429f261fe165d3c6bc4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR