Pyridoxine hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
Adermine hydrochloride, Vitamin B6 hydrochloride, PN HCl, Pyridoxine HCl, (5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol hydrochloride, Pyridoxol hydrochloride, Pyridoxine HCI
CAS Number: 58-56-0
MDL Number: MFCD00012807
Molecular Formula: C8H11NO3
Molecular Weight: 205.64
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 207
Product Categories: Bioscience, Vitamins
Other Product Names / Synonyms: Adermin Hydrochloride, Vitamin B6 Hydrochloride
Formula:
C₈H₁₂ClNO₃ MW: 205.64 g/mol Melting Pt: 207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012807 CAS Number: 58-56-0 EINECS: 200-386-2 Merck Index: 13,08072 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...