Pyridine-N-oxide ≥98.0%

Supplier: TCI America

Synonyms: Pyridine 1-oxide, Pyridine N-oxide

P0557-500G P0557-25G P0557-100G
TCP0557-500GEA 489.8 CAD
TCP0557-500G TCP0557-025G TCP0557-100G
Pyridine-N-oxide ≥98.0%
Pyridine N-oxide

CAS Number: 694-59-7
MDL Number: MFCD00006194
Molecular Formula: C5H5NO
Molecular Weight: 95.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 105
Melting point (°C): 62

Formula: C₅H₅NO
Boiling Pt: 105 °C
Melting Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006194
CAS Number: 694-59-7
EINECS: 211-774-6
Merck Index: 12,08155
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcc00bd76771a70b49cef7404dd78b68ce05ca6c&vid=bcc00bd76771a70b49cef7404dd78b68ce05ca6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR