Pyridine-2,6-dicarboxylic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2,6-Pyridinedicarboxylic acid, Dipicolinic acid, DPA

P0554-25G P0554-500G P0554-100G
TCP0554-025GEA 56.25 CAD
TCP0554-025G TCP0554-500G TCP0554-100G
Pyridine-2,6-dicarboxylic acid ≥98.0%
Pyridine-2,6-dicarboxylic acid

CAS Number: 499-83-2
MDL Number: MFCD00006299
Molecular Formula: C7H5NO4
Molecular Weight: 167.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 252
Flash Point (°C): 188


Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents, Monomers
Other Product Names / Synonyms: Dipicolinic Acid

Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 357
Melting Pt: 327
Flash Pt: 188
Storage Temperature: Ambient
MDL Number: MFCD00006299
CAS Number: 499-83-2
EINECS: 207-894-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=113734767f845c8e82fbb084f4189bf2c50631f2&vid=113734767f845c8e82fbb084f4189bf2c50631f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR