Pyridine-3,5-dicarboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

P0551-5G P0551-25G
TCP0551-005GEA 26.86 CAD
TCP0551-005G TCP0551-025G
Pyridine-3,5-dicarboxylic acid ≥98.0% (by HPLC, titration analysis)
Pyridine-3,5-dicarboxylic acid

CAS Number: 499-81-0
MDL Number: MFCD00006393
Molecular Formula: C7H5NO4
Molecular Weight: 167.12
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₇H₅NO₄
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 320…325 °C
Storage Temperature: Ambient
MDL Number: MFCD00006393
CAS Number: 499-81-0
EINECS: 207-893-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9ee710b206bd8f1563a161c7d80eb973a792971&vid=b9ee710b206bd8f1563a161c7d80eb973a792971&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR