Pyridine-2,3-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
P0550-500G
P0550-25G
TCP0550-500GEA
295.93
CAD
TCP0550-500G
TCP0550-025G
Pyridine-2,3-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Pyridine-2,3-dicarboxylic acid
CAS Number: 89-00-9
MDL Number: MFCD00006295
Molecular Formula: C7H5NO4
Molecular Weight: 167.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 188
Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents, Monomers
Other Product Names / Synonyms: Quinolinic Acid
Formula:
C₇H₅NO₄ Boiling Pt: 357 °C (1013 hPa) Melting Pt: 188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006295 CAS Number: 89-00-9 EINECS: 201-874-8 Merck Index: 12,08258 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...