Propiophenone ≥98.0%

Supplier: TCI America

Synonyms: Phenyl ethyl ketone, Ethyl phenyl ketone, 1-Phenyl-1-propanone

P0519-25G P0519-500G
TCP0519-025GEA 51.38 CAD
TCP0519-025G TCP0519-500G
Propiophenone ≥98.0%
Propiophenone

CAS Number: 93-55-0
MDL Number: MFCD00009309
Molecular Formula: C9H10O
Molecular Weight: 134.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 216
Flash Point (°C): 92
Freezing point (°C): 18
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: Ethyl Phenyl Ketone, Propionylbenzene

Formula: C₆H₅C(O)CH₂CH₃
Boiling Pt: 216 °C
Melting Pt: 18 °C
Density: 1.01 (20 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00009309
CAS Number: 93-55-0
EINECS: 202-257-6
Merck Index: 12,08015
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e34c7b8eaf3ea974e70e8eace2c8b9aec54ad16&vid=6b115d9126ef33a29e0f649597246f00db797dad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR