Calcium propionate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Calcium dipropionate, Propionic acid hemicalcium salt, Propionic acid calcium salt
P0503-25G
P0503-500G
TCP0503-025GEA
25.72
CAD
TCP0503-025G
TCP0503-500G
Calcium propionate ≥98.0% (by titrimetric analysis)
Calcium propionate
CAS Number: 4075-81-4
MDL Number: MFCD00036137
Molecular Formula: C3H6O2
Molecular Weight: 186.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: Propionic Acid Calcium Salt
Formula:
(CH₃CH₂COO)₂Ca MW: 186.22 g/mol Melting Pt: > 300 °C |
MDL Number:
MFCD00036137 CAS Number: 4075-81-4 EINECS: 223-795-8 Merck Index: 13,01703 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...