Tricarballylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

P0490-25G P0490-100G
TCP0490-025GEA 205.4 CAD
TCP0490-025G TCP0490-100G
Tricarballylic acid ≥98.0% (by GC, titration analysis)
Tricarballylic acid

CAS Number: 99-14-9
MDL Number: MFCD00002723
Molecular Formula: C6H8O6
Molecular Weight: 176.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 159


Other Product Names / Synonyms: Tricarballylic Acid

Formula: C₆H₈O₆
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 159 °C
Storage Temperature: Ambient
MDL Number: MFCD00002723
CAS Number: 99-14-9
EINECS: 202-733-3
Merck Index: 12,09752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fdd3c56040008cb679d8aff9115581d5c13ed96&vid=4fdd3c56040008cb679d8aff9115581d5c13ed96&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR