1,3-Propanediol ≥98.0%

Supplier: TCI America

P0486-500G P0486-25G P0486-100G
TCP0486-500GEA 164.1 CAD
TCP0486-500G TCP0486-025G TCP0486-100G
1,3-Propanediol ≥98.0%

CAS Number: 504-63-2
MDL Number: MFCD00002949
Molecular Formula: C3H8O2
Molecular Weight: 76.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 214
Flash Point (°C): 74
Specific Gravity (20/20): 1.05


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,3-Dihydroxypropane, Trimethylene Glycol

Formula: HO(CH₂)₃OH
Boiling Pt: 214 °C
Melting Pt: –32 °C
Density: 1.05 (20 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00002949
CAS Number: 504-63-2
EINECS: 207-997-3
Merck Index: 12,09845
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a7363955f07cab3bfef1fd43027d3aa42893b8a&vid=3f075951cc0c100134ee17a06cca970f7421d13e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR