L(-)-Proline ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

P0481-250G P0481-25G
TCP0481-250GEA 191.43 CAD
TCP0481-250G TCP0481-025G
L(-)-Proline ≥99.0% (by HPLC, titration analysis)
L(-)-Proline

CAS Number: 147-85-3
MDL Number: MFCD00064318
Molecular Formula: C5H9NO2
Molecular Weight: 115.13
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 228
Specific rotation [a]20/D: -85 deg (C=4, H2O)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Bioscience, Regenerative Medicine Research Reagents, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Pro-OH

Formula: C₅H₉NO₂
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 228 °C
Density: 1.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064318
CAS Number: 147-85-3
EINECS: 205-702-2
Merck Index: 13,07871
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b40e836604d92b1c16dfcfacf93fd34e26cb8eac&vid=b40e836604d92b1c16dfcfacf93fd34e26cb8eac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR