DL-Proline ≥99.0%

Supplier: TCI America

Synonyms: (±)-Proline, H-DL-Pro-OH

P0480-1G P0480-25G P0480-5G
TCP0480-001GEA 39.5 CAD
TCP0480-001G TCP0480-025G TCP0480-5G
DL-Proline ≥99.0%
DL-Proline

CAS Number: 609-36-9
MDL Number: MFCD00005250
Molecular Formula: C5H9NO2
Molecular Weight: 115.13
Purity/Analysis Method: >99.0% (T)
Form: Crystal


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-DL-Pro-OH

Formula: C₅H₉NO₂
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 208…210 °C
Density: 1.38 g/cm³ (22.9 °C)
MDL Number: MFCD00005250
CAS Number: 609-36-9
EINECS: 210-189-3
Merck Index: 12,07963
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8325448b726b813f96a1e8e734dab4cd303b1298&vid=8325448b726b813f96a1e8e734dab4cd303b1298&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR