Polyvinylpyrrolidone

Supplier: TCI America

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

P0471-25G P0471-500G P0471-100G
TCP0471-025GEA 51.08 CAD
TCP0471-025G TCP0471-500G TCP0471-100G
Polyvinylpyrrolidone
Polyvinylpyrrolidone

CAS Number: 9003-39-8
Form: Powder
Color: White
Flash Point (°C): 215


Product Categories: Materials Chemistry, Polymer Research Reagents

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD00149016
CAS Number: 9003-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR