Piperonyl alcohol ≥98.0%

Supplier: TCI America

P0457-500G P0457-25G P0457-100G
TCP0457-500GEA 759.98 CAD
TCP0457-500G TCP0457-025G TCP0457-100G
Piperonyl alcohol ≥98.0%
Piperonyl alcohol

CAS Number: 495-76-1
MDL Number: MFCD00005836
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 157
Melting point (°C): 54
Flash Point (°C): 110


Other Product Names / Synonyms: (1,3-Benzodioxol-5-yl)methanol, Helioalcohol, 3,4-Methylenedioxybenzyl Alcohol

Formula: C₈H₈O₃
Boiling Pt: 157 °C
Melting Pt: 54 °C
Flash Pt: 110 °C
MDL Number: MFCD00005836
CAS Number: 495-76-1
EINECS: 207-808-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3d5e1289dfd9a2fcaf89496ee30555d08783242&vid=d3d5e1289dfd9a2fcaf89496ee30555d08783242&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR