2-Piperidone ≥98.0%
Supplier: TCI America
|
Warning
|
P0455-25G
P0455-500G
P0455-100G
TCP0455-025GEA
109.02
CAD
TCP0455-025G
TCP0455-500G
TCP0455-100G
2-Piperidone ≥98.0%
2-Piperidone
CAS Number: 675-20-7
MDL Number: MFCD00006037
Molecular Formula: C5H9NO
Molecular Weight: 99.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 256
Flash Point (°C): 147
Freezing point (°C): 38
Other Product Names / Synonyms: 2-Azacyclohexanone, delta-Valerolactam
Formula:
C₅H₉NO Boiling Pt: 256 °C Melting Pt: 35…38 °C Density: 1.073 g/cm³ (40 °C) Flash Pt: 147 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006037 CAS Number: 675-20-7 EINECS: 211-622-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...