Picolinic acid ≥99.0%

Supplier: TCI America
Warning

Synonyms: Pyridine-2-carboxylic acid, 2-Picolinic acid, 2-Pyridinecarboxylic acid

P0421-25G P0421-500G P0421-100G
TCP0421-025GEA 36.34 CAD
TCP0421-025G TCP0421-500G TCP0421-100G
Picolinic acid ≥99.0%
Picolinic acid

CAS Number: 98-98-6
MDL Number: MFCD00006293
Molecular Formula: C6H5NO2
Molecular Weight: 123.11
Purity/Analysis Method: >99.0% (T)
Form: Crystalline Powder
Melting point (°C): 140


Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents, Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: Picolinic Acid

Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 293
Melting Pt: 140 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006293
CAS Number: 98-98-6
EINECS: 202-719-7
Merck Index: 12,07557
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f2410f3c7c2b8e4f5a03d34b2b36be8ae35f3ef&vid=6f2410f3c7c2b8e4f5a03d34b2b36be8ae35f3ef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR