2-Picoline-N-oxide 99,0

Supplier: TCI America

Synonyms: 2-Methylpyridine-N-oxide, 2-Picoline N-oxide, 2-Methylpyridine-1-oxide, 2-Methylpyridine 1-oxide

P0418-500G P0418-25G
TCP0418-500GEA 397.12 CAD
TCP0418-500G TCP0418-025G
2-Picoline-N-oxide 99,0
2-Picoline N-oxide

CAS Number: 931-19-1
MDL Number: MFCD00006199
Molecular Formula: C6H7NO
Molecular Weight: 109.13
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 261
Flash Point (°C): 143
Freezing point (°C): 48


Other Product Names / Synonyms: 2-Picoline N-Oxide

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 259
Melting Pt: 0
Flash Pt: 143 °C
MDL Number: MFCD00006199
CAS Number: 931-19-1
EINECS: 213-230-3

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR