Phthalimide potassium salt ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P0403-500G P0403-25G
TCP0403-500GEA 121.66 CAD
TCP0403-500G TCP0403-025G
Phthalimide potassium salt ≥98.0% (by titrimetric analysis)
Phthalimide potassium salt

CAS Number: 1074-82-4
MDL Number: MFCD00005887
Molecular Formula: C8H5NO2
Molecular Weight: 185.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₈H₄KNO₂
Melting Pt: > 300 °C
Density: 1.634 g/cm³ (20 °C)
MDL Number: MFCD00005887
CAS Number: 1074-82-4
EINECS: 214-046-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=374222e739ae1c177e07378f0eb76072bec96029&vid=374222e739ae1c177e07378f0eb76072bec96029&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR