Phthalimide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P0402-500G P0402-25G
TCP0402-500GEA 72.96 CAD
TCP0402-500G TCP0402-025G
Phthalimide ≥98.0% (by GC, titration analysis)
Phthalimide

CAS Number: 85-41-6
MDL Number: MFCD00005881
Molecular Formula: C8H5NO2
Molecular Weight: 147.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 234
Flash Point (°C): 150

Formula: C₈H₅NO₂
Boiling Pt: 366 °C (1013 hPa)
Melting Pt: 234 °C
Density: 1.21 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00005881
CAS Number: 85-41-6
EINECS: 201-603-3
Merck Index: 12,07529
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d40eeeeca3251e67c3a81f1484d60e083ece988a&vid=d40eeeeca3251e67c3a81f1484d60e083ece988a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR