Phthalide ≥98.0%

Supplier: TCI America
Warning

P0401-25G P0401-500G
TCP0401-025GEA 26.86 CAD
TCP0401-025G TCP0401-500G
Phthalide ≥98.0%
Phthalide

CAS Number: 87-41-2
MDL Number: MFCD00005906
Molecular Formula: C8H6O2
Molecular Weight: 134.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 75
Flash Point (°C): 135

Formula: C₈H₆O₂
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 75 °C
Density: 1.1 g/cm³ (30 °C)
Flash Pt: 135 °C
MDL Number: MFCD00005906
CAS Number: 87-41-2
EINECS: 201-744-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfe7e5390c218911ebafbb70c5fea325a8d66e58&vid=dfe7e5390c218911ebafbb70c5fea325a8d66e58&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR