2-Piperidinoethanol ≥98.0%

Supplier: TCI America

P0348-500G P0348-25G
TCP0348-500GEA 151.68 CAD
TCP0348-500G TCP0348-025G
2-Piperidinoethanol ≥98.0%
2-Piperidinoethanol

CAS Number: 3040-44-6
MDL Number: MFCD00006512
Molecular Formula: C7H15NO
Molecular Weight: 129.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 198
Flash Point (°C): 73
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: 1-(2-Hydroxyethyl)piperidine

Formula: C₇H₁₅NO
Boiling Pt: 198 °C
Density: 0.98 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00006512
CAS Number: 3040-44-6
EINECS: 221-244-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1615ad9d01d161f4f4e5fdf47843c0b441500977&vid=1615ad9d01d161f4f4e5fdf47843c0b441500977&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR