1-Pentanesulfonic acid sodium salt ≥98.0%
Supplier: TCI America
Synonyms:
Sodium 1-pentanesulphonate, Sodium pentane-1-sulphonate, Pentane-1-sulphonic acid sodium salt, Sodium pentanesulphonate, 98%, Sodium pentanesulphonate, Pentane-1-sulfonic acid sodium salt, Sodium pentanesulfonate, Sodium 1-pentanesulfonate, Sodium pentane-1-sulfonate, Sodium pentanesulfonate, 98%
CAS Number: 22767-49-3
MDL Number: MFCD00007541
Molecular Formula: C5H12O3S
Molecular Weight: 174.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: Amylsulfonic Acid Sodium Salt, 1-Pentanesulfonic Acid Sodium Salt, Sodium Amylsulfonate
Formula:
C₅H₁₁O₃SNa MW: 174.2 g/mol Melting Pt: > 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007541 CAS Number: 22767-49-3 EINECS: 245-208-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...