Diethyl phenylphosphonate ≥96.0%
Supplier: TCI America
P0335-25ML
P0335-5ML
TCP0335-025MLEA
474
CAD
TCP0335-025ML
TCP0335-5ML
Diethyl phenylphosphonate ≥96.0%
Diethyl phenylphosphonate
CAS Number: 1754-49-0
MDL Number: MFCD00015136
Molecular Formula: C10H15O3P
Molecular Weight: 214.20
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 267
Specific Gravity (20/20): 1.12
Other Product Names / Synonyms: Benzenephosphonic Acid Diethyl Ester, Diethyl Benzenephosphonate, Phenylphosphonic Acid Diethyl Ester
Formula:
C₁₀H₁₅O₃P Boiling Pt: 267 °C Density: 1.12 (20 °C) |
MDL Number:
MFCD00015136 CAS Number: 1754-49-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...