Diethyl phenylmalonate ≥97.0%
Supplier: TCI America
P0311-25ML
P0311-500ML
TCP0311-025MLEA
30.56
CAD
TCP0311-025ML
TCP0311-500ML
Diethyl phenylmalonate ≥97.0%
Diethyl phenylmalonate
CAS Number: 83-13-6
MDL Number: MFCD00009144
Molecular Formula: C13H16O4
Molecular Weight: 236.27
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 285
Flash Point (°C): 113
Freezing point (°C): 17
Specific Gravity (20/20): 1.10
Other Product Names / Synonyms: Phenylmalonic Acid Diethyl Ester
Formula:
C₁₃H₁₆O₄ Boiling Pt: 285 °C Density: 1.10 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00009144 CAS Number: 83-13-6 EINECS: 201-456-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...