Potassium hydrogen phthalate ≥99.8%

Supplier: TCI America

Synonyms: Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt

P0309-25G P0309-500G
TCP0309-025GEA 45.91 CAD
TCP0309-025G TCP0309-500G
Potassium hydrogen phthalate ≥99.8%
Potassium hydrogen phthalate

CAS Number: 877-24-7
MDL Number: MFCD00013070
Molecular Formula: C8H6O4
Molecular Weight: 204.22
Purity/Analysis Method: >99.8% (T)
Form: Crystal


Product Categories: Bioscience, Buffer-related Reagents
Other Product Names / Synonyms: Monopotassium Phthalate, Phthalic Acid Monopotassium Salt

Formula: HOOCC₆H₄COOK
MW: 204.22 g/mol
Melting Pt: 295…300 °C
Density: 1.6362 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013070
CAS Number: 877-24-7
EINECS: 212-889-4
Merck Index: 13,07694
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33dc082c9c7caaa1309254cdff228673bfd03d23&vid=33dc082c9c7caaa1309254cdff228673bfd03d23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.8% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR