Diphenyl phthalate ≥98.0%
Supplier: TCI America
|
Warning
|
P0305-25G
P0305-500G
TCP0305-025GEA
42.15
CAD
TCP0305-025G
TCP0305-500G
Diphenyl phthalate ≥98.0%
Diphenyl phthalate
CAS Number: 84-62-8
MDL Number: MFCD00003038
Molecular Formula: C20H14O4
Molecular Weight: 318.33
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 257
Melting point (°C): 75
Flash Point (°C): 224
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Phthalic Acid Diphenyl Ester
Formula:
C₂₀H₁₄O₄ Boiling Pt: 257 °C Melting Pt: 75 °C Density: 1.313 g/cm³ (20 °C) Flash Pt: 224 °C |
MDL Number:
MFCD00003038 CAS Number: 84-62-8 EINECS: 201-546-4 Merck Index: 12,07460 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...